Läs mer
Ammonium Geranyl Pyrophosphate Triammonium Salt, TRC
Produktkod. 30334597
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30334597 1 mg
1 options
Produktkod. 30334597 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6340151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ammonium Geranyl Pyrophosphate triammonium salt
CAS 116057-55-7
Molekylformel C10 H20 O7 P2 . 3 H3 N
Kvantitet 1 mg
Synonym Diphosphoric Acid Mono[(3E)-3,7-dimethyl-2,6-octadienyl] Ester Ammonium Salt (1:3)
InChI formel InChI=1S/C10H20O7P2.3H3N/c1-9(2)5-4-6-10(3)7-8-16-19(14,15)17-18(11,12)13;;;/h5,7H,4,6,8H2,1-3H3,(H,14,15)(H2,11,12,13);3*1H3/b10-7+;;;
LEDER N.N.N.CC(=CCC\C(=C\COP(=O)(O)OP(=O)(O)O)\C)C
IUPAC-namn azane;[(2E)-3,7-dimethylocta-2,6-dienyl] phosphono hydrogen phosphate
Molekylvikt (g/mol) 365.3
Formel vikt 365.1481