missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydrogen Carbonate, Certified AR for analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
1 kg
Description
Ammonium Hydrogen Carbonate, Certified AR for analysis
Specifications
Melting Point | 105°C |
Color | White |
pH | 8 |
Physical Form | Powder/Solid |
Quantity | 250 g |
Formula Weight | 79.06g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 67 hPa at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'