missing translation for 'onlineSavingsMsg'
Läs mer

Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals

Produktkod. p-7051850
Klicka för att se tillgängliga alternativ
Kvantitet:
50 g
250 g
Förpackningsstorlek:
250g
50g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11389767

Brand: Thermo Scientific Alfa Aesar 042112.18

Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

In photography, blueprints

In photography, blueprints

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 13268-42-3
Molekylformel C6H18FeN3O15
Molekylvikt (g/mol) 428.06
MDL-nummer MFCD00150137
InChI-nyckel ARJSTCBPHIPXIJ-UHFFFAOYSA-K
Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
IUPAC-namn triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
LEDER [NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4

Specifikationer

Kemiskt namn eller material Ammonium iron(III) oxalate hydrate
Smältpunkt (decomposition)
Kvantitet 50 g
UN-nummer UN3288
Känslighet Hygroscopic; Light sensitive
Merck Index 14,517
Löslighetsinformation Very soluble in water; Insoluble in alcohol
Formel vikt 374.02 (Anhydrous)
Fysisk form Powder
compliance-icons
Produktbeteckning
  • Ammonium iron(III) oxalate
Signalord
  • Varning
Riskkategorier
  • Akut toxicitet Kategorija 4
Hazard Statement
  • H302-Skadligt vid förtäring.
  • H312-Skadligt vid hudkontakt.
Precautionary Statement
  • P280-Använd skyddshandskar/skyddskläder/ögonskydd/ansiktsskydd.
  • P301+P312-VID FÖRTÄRING: Kontakta GIFTINFORMATIONSCENTRAL / läkare / /om du mår dåligt.
  • P363-Nedstänkta kläder ska tvättas innan de används igen.
  • P321-Särskild behandling (se på etiketten).
  • P501-Innehållet/behållaren lämnas till .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.