Läs mer
Ammonium Molybdate Solution R6 - British Pharmacopoeia, Fisher Chemical™
Handla allt Fisher Chemical ProdukterBeskrivning
- This Pharmacopoeia reagent is manufactured :
- Using pharmacopoeia reagent quality components
- Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
- Supplied with a Certificate of Analysis for Performance Test
- Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034
Hazard Statement: May be flammable or oxidizing; may cause severe skin burns and serious eye damage or irritation; may cause respiratory irritation, drowsiness or dizziness; may be toxic if swallowed, in contact with skin, or if inhaled; may cause cancer, genetic defects, reproductive toxicity or developmental harm; may be harmful to aquatic life with long-lasting effects; may be corrosive to metals.
Specifikationer
Specifikationer
| Kvantitet | 100 mL |