Läs mer
Ammonium Nonanoate, TRC
Produktkod. 30332472
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30332472 500 mg
1 options
Produktkod. 30332472 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA633695500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nonanoic Acid Ammonium
CAS 63718-65-0
Molekylformel C9 H17 O2 . H4 N
Kvantitet 500 mg
Synonym Ammonium Nonanoate, Nonanoic acid, ammonium salt (9CI), Ammonium nonanoate, Ammonium pelargonate, Pelargonic acid ammonium salt, Nonanoic Acid Ammonium
InChI formel InChI=1S/C9H18O2.H3N/c1-2-3-4-5-6-7-8-9(10)11;/h2-8H2,1H3,(H,10,11);1H3
LEDER O=C(O)CCCCCCCC.N
IUPAC-namn ammonia;nonanoic acid
Molekylvikt (g/mol) 175.268
Formel vikt 175.157