Läs mer
Ammonium Oxalate, TRC
Produktkod. 30348382
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30348382 1 g
1 options
Produktkod. 30348382 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6341301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ammonium Oxalate
CAS 1113-38-8
Molekylformel C2H2O4 . 2NH3
Kvantitet 1 g
Synonym Diammonium Oxalate
InChI formel InChI=1S/C2H2O4.2H3N/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H3
LEDER [NH4+].[NH4+].[O-]C(=O)C(=O)[O-]
IUPAC-namn diazanium;oxalate
Molekylvikt (g/mol) 124.1
Formel vikt 124.0484