Läs mer
Ammonium Perfluoro(2-methyl-3-oxahexanoate), TRC
Produktkod. 30336844
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30336844 50 mg
1 options
Produktkod. 30336844 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63496050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Perfluoro-2-methyl-3-oxahexanoate ammonium salt
CAS 62037-80-3
Molekylformel C6 F11 O3 . H4 N
Kvantitet 50 mg
Synonym Propanoic acid, 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)-, ammonium salt (1:1), Propanoic acid, 2,3,3,3-tetrafluoro-2-(heptafluoropropoxy)-, ammonium salt (9CI), Ammonium 2-(perfluoropropoxy)perfluoropropionate, Ammonium perfluoro-2-methyl-3-oxahexanoate
InChI formel InChI=1S/C6HF11O3.H3N/c7-2(1(18)19,4(10,11)12)20-6(16,17)3(8,9)5(13,14)15;/h(H,18,19);1H3
LEDER [NH4+].[O-]C(=O)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F
IUPAC-namn azanium;2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoate
Molekylvikt (g/mol) 347.08
Formel vikt 347.0016