missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072832
Click to view available options
Quantity:
1 kg
3 kg
This item is not returnable. View return policy
7727-54-0
H8N2O8S2
228.19
3390
ROOXNKNUYICQNP-UHFFFAOYSA-N
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
This item is not returnable. View return policy
7727-54-0
H8N2O8S2
228.19
3390
ROOXNKNUYICQNP-UHFFFAOYSA-N
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Melting Point 120°C
Color White
pH 3.2
Physical Form Powder/Solid
Quantity 1 kg
Formula Weight 228.19g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'