Läs mer
Ammonium Polyphosphate, TRC
Produktkod. 30340259
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30340259 500 mg
1 options
Produktkod. 30340259 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA634983500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ammonium Polyphosphate
CAS 68333-79-9
Molekylformel (NH4PO3)n
Kvantitet 500 mg
Synonym Ammonium Polyphosphate
InChI formel InChI=1S/3H3N.H3O4P/c;;;1-5(2,3)4/h3*1H3;(H3,1,2,3,4)
LEDER [NH4+].[NH4+].[NH4+].[O-]P(=O)([O-])[O-]
IUPAC-namn triazanium;phosphate
Molekylvikt (g/mol) 149.09
Formel vikt 149.06