missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Purpurate, Pure, Metal Indicator, Fisher Chemical™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 300°C |
Color | Red |
Physical Form | Powder/Solid |
Quantity | 5 g |
Formula Weight | 284.19g/mol |
Packaging | Glass Jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'