Läs mer
Ammonium Standard: NH4+ @ 100 μg/mL in H2O, VHG™
Produktkod. 17998234
Change view
Kvantitet:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17998234 500 ml
17908534 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17998234 Leverantör VHG Labs Leverantörsnummer VHGINH4100500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Ammonium Standard: NH4+ @ 100 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ammonium
CAS 14798-03-9
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnamn Ammonium
Frakt skick Room Temperature
Molekylformel NH4+
Synonym NH4+
LEDER [NH4+]
IUPAC-namn Ammonium
Lösningsmedel eller matris Water
Molekylvikt (g/mol) 18.04
Kvantitet 500 ml
Formel vikt 18.04 g/mol