missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tartrate, 98.8%, MP Biomedicals™

Catalog Number p-4405771
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
3164-29-2
C4H12N2O6
184.15
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
This item is not returnable. View return policy
3164-29-2
C4H12N2O6
184.15
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 0.988
Formula Weight 184.1
Chemical Name or Material Ammonium tartrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'