missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, Honeywell Fluka™

Catalog Number 15613250
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
6 x 500 g
40 kg
6 x 1 kg
This item is not returnable. View return policy
1762-95-4
CH4N2S
76.117
MFCD00011428
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
15666
azanium;thiocyanate
C(#N)[S-].[NH4+]
This item is not returnable. View return policy
1762-95-4
CH4N2S
76.117
MFCD00011428
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
15666
azanium;thiocyanate
C(#N)[S-].[NH4+]

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99% (AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 6 x 1 kg
Linear Formula NH4SCN
Formula Weight 76.12
Percent Purity ≥99% (AT)
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.02% (as SO4)
Chemical Name or Material Ammonium Thiocyanate, Reag. Ph. Eur. reag. ISO
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'