missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
6 x 500 g
40 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 40 kg |
Linear Formula | NH4SCN |
Beilstein | 3595135 |
Formula Weight | 76.12g/mol |
Percent Purity | ≥99% (AT) |
Grade | ACS/puriss. p.a. |
Residue after Ignition | ≤0.02% (as SO4) |
Chemical Name or Material | Ammonium Thiocyanate, Reag. Ph. Eur. reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'