missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14694-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
AMN1 belongs to the AMN1 family. The exact function of AMN1 remains unknown.Specifications
AMN1 | |
Polyclonal | |
Unconjugated | |
AMN1 | |
AMN1, Protein AMN1 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
196394, 232566, 302032 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5U201, Q8IY45 | |
AMN1 | |
AMN1 Fusion Protein Ag6257 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |