missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | AMN1 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823464
|
Proteintech
14694-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813464
|
Proteintech
14694-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AMN1 belongs to the AMN1 family. The exact function of AMN1 remains unknown.Specifications
AMN1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q5U201, Q8IY45 | |
196394, 232566, 302032 | |
AMN1 Fusion Protein Ag6257 | |
Primary | |
-20°C | |
AMN1 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AMN1, Protein AMN1 homolog | |
AMN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |