Läs mer
Amolanone Hydrochloride, LoGiCal
Produktkod. 30458142
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30458142 5 mg
1 options
Produktkod. 30458142 Leverantör LoGiCal Leverantörsnummer LGCFOR1775.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amolanone Hydrochloride
CAS 6009-67-2
Analyt- eller komponentnamn Amolanone Hydrochloride
Frakt skick Room Temperature
Molekylformel C20 H23 N O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym 2(3H)-Benzofuranone, 3-[2-(diethylamino)ethyl]-3-phenyl-, hydrochloride (8CI,9CI), Amethone Hydrochloride, Amolanone hydrochloride, 2(3H)-Benzofuranone, 3-[2-(diethylamino)ethyl]-3-phenyl-, hydrochloride (1:1)
InChI formel InChI=1S/C20H23NO2.ClH/c1-3-21(4-2)15-14-20(16-10-6-5-7-11-16)17-12-8-9-13-18(17)23-19(20)22;/h5-13H,3-4,14-15H2,1-2H3;1H
Lösningstyp Neat
LEDER Cl.CCN(CC)CCC1(C(=O)Oc2ccccc12)c3ccccc3
IUPAC-namn 3-[2-(diethylamino)ethyl]-3-phenyl-1-benzofuran-2-one;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 345.86
Kvantitet 5 mg
Formel vikt 345.1496 g/mol