Läs mer
Amopyroquine, TRC
Produktkod. 30333063
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30333063 5 mg
1 options
Produktkod. 30333063 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6342155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amopyroquine
CAS 550-81-2
Molekylformel C20 H20 Cl N3 O
Kvantitet 5 mg
Synonym o-Cresol, 4-[(7-chloro-4-quinolyl)amino]-α-1-pyrrolidinyl- (6CI,7CI,8CI), 4-[(7-Chloro-4-quinolyl)amino]-α-1-pyrrolidinyl-o-cresol, Amopyroquin, Amopyroquine, PAM-780, Propoquin, Phenol, 4-[(7-chloro-4-quinolinyl)amino]-2-(1-pyrrolidinylmethyl)
InChI formel InChI=1S/C20H20ClN3O/c21-15-3-5-17-18(7-8-22-19(17)12-15)23-16-4-6-20(25)14(11-16)13-24-9-1-2-10-24/h3-8,11-12,25H,1-2,9-10,13H2,(H,22,23)
LEDER Oc1ccc(Nc2ccnc3cc(Cl)ccc23)cc1CN4CCCC4
IUPAC-namn 4-[(7-chloroquinolin-4-yl)amino]-2-(pyrrolidin-1-ylmethyl)phenol
Molekylvikt (g/mol) 353.85
Formel vikt 353.1295