Läs mer
Ampeloptin, TRC
Produktkod. 30338563
Change view
Kvantitet:
1 g
Product Code. Kvantitet
30338563 1 g
1 options
Product Code. 30338563 Supplier Toronto Research Chemicals Supplier No. TRCA6348001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Dihydromyricetin
CAS 27200-12-0
Molekylformel C15 H12 O8
Kvantitet 1 g
Synonym 4H-1-Benzopyran-4-one, 2,3-dihydro-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-, (2R,3R)-, 4H-1-Benzopyran-4-one, 2,3-dihydro-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-, (2R-trans)-, Ampelopsin (6CI), Flavanone, 3,3',4',5,5',7-hexahydroxy- (7CI,8CI), (+)-Ampelopsin, (+)-Dihydromyricetin, (2R,3R)-2,3-Dihydro-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-1-benzopyran-4-one, Ampelopsin (flavanol), Ampeloptin, Dihydromyricetin
InChI formel InChI=1S/C15H12O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,14-20,22H/t14-,15+/m0/s1
LEDER O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3cc(O)c(O)c(O)c3
IUPAC-namn (2R,3R)-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one
Molekylvikt (g/mol) 320.25
Formel vikt 320.0532