Läs mer
Amperozide Dihydrochloride, TRC
Produktkod. 30331519
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30331519 10 mg
1 options
Produktkod. 30331519 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63480310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amperozide Dihydrochloride
CAS 1307807-06-2
Molekylformel C23H29F2N3O . 2(HCl)
Kvantitet 10 mg
Synonym 4-[4,4-bis(4-Fluorophenyl)butyl]-N-ethyl-1-piperazinecarboxamide Dihydrochloride, 4-[4,4-bis(4-Fluorophenyl)butyl]-N-ethyl-1-piperazinecarboxamide Hydrochloride (1:2)
InChI formel InChI=1S/C23H29F2N3O.2ClH/c1-2-26-23(29)28-16-14-27(15-17-28)13-3-4-22(18-5-9-20(24)10-6-18)19-7-11-21(25)12-8-19;;/h5-12,22H,2-4,13-17H2,1H3,(H,26,29);2*1H
LEDER FC1=CC=C(C(C2=CC=C(F)C=C2)CCCN3CCN(C(NCC)=O)CC3)C=C1.Cl.Cl
IUPAC-namn 4-(4,4-bis(4-fluorophenyl)butyl)-N-ethylpiperazine-1-carboxamide Dihydrochloride
Molekylvikt (g/mol) 474.41
Formel vikt 473.19