missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphiphysin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13379-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Amphiphysins, members of the BAR (Bin-Amphiphysin-Rvsp) protein super family, play a key role in clathrin-mediated endocytosis of synaptic vesicles (SVs). All members of the BAR family share a highly conserved N-terminal BAR domain and a C-terminal Src homology (SH3) domain. Two isoforms of amphiphysin have been identified and act in concert as a heterodimer. Amphiphysins interact via its carboxy-terminal SH3 domain with dynamin, synaptojanin and clathrin. These complexes are implicated in synaptic vesicle recycling at nerve terminal. Amphiphysins were also identified as the domit autoantigen in paraneoplastic Stiff-Man Syndrome.Specifications
Amphiphysin | |
Polyclonal | |
Unconjugated | |
AMPH | |
AMPH, AMPH1, Amphiphysin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
218038, 273, 60668 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08838, P49418, Q7TQF7 | |
Amph | |
Amphiphysin Fusion Protein Ag4204 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |