Läs mer
Amphiregulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15379
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Amphiregulin Polyclonal specifically detects Amphiregulin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| Western Blot, Immunohistochemistry (Paraffin) | |
| Rabbit | |
| 0.1 mL | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Polyclonal | |
| Recombinant protein encompassing a sequence within the center region of human Amphiregulin. The exact sequence is proprietary. | |
| RUO | |
| IgG |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering