Läs mer
Ampicillin Desoxocarboxylic Acid-[(R)-Phenylglycine], TRC
Produktkod. 30338151
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30338151 5 mg
1 options
Produktkod. 30338151 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6343555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ampicillin desoxocarboxylic acid-[(R)-Phenylglycine]
CAS 1207726-28-0
Molekylformel C24 H26 N4 O5 S
Kvantitet 5 mg
Synonym (αR)-α-[[[(2S,5R,6R)-6-[[(2R)-2-Amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]amino]-benzeneacetic Acid
InChI formel InChI=1S/C24H26N4O5S/c1-24(2)18(20(30)26-16(23(32)33)14-11-7-4-8-12-14)28-21(31)17(22(28)34-24)27-19(29)15(25)13-9-5-3-6-10-13/h3-12,15-18,22H,25H2,1-2H3,(H,26,30)(H,27,29)(H,32,33)/t15-,16-,17-,18+,22-/m1/s1
LEDER CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)N[C@@H](C(=O)O)c4ccccc4
IUPAC-namn (2R)-2-[[(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]amino]-2-phenylacetic acid
Molekylvikt (g/mol) 482.55
Formel vikt 482.1624