Learn More
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
Product Code. 10419313
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Poly Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10419313 25 g Poly Bottle 25g
10193433 5 g Poly Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10419313 Supplier Fisher BioReagents Supplier No. BP176025
 Restricted Item
I lager
Add to basket
For selection for ampicillin resistance in mutated and transformed cells | CAS: 69-52-3 | C16H21N3NaO4S | 374.411 g/mol
For selection for ampicillin resistance in mutated and transformed cells
For selection for ampicillin resistance in mutated and transformed cells. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69-52-3
Molecular Formula C16H21N3NaO4S
Molecular Weight (g/mol) 374.411
InChI Key BSFVNXCYXDYHOD-ZQDFAFASSA-N
Synonym Ampicillin
PubChem CID 131673879
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]
Specifications
Type Ampicillin Sodium Salt
Physical Form Solid
pH 8.0 to 10.0
Melting Point 215°C
Packaging Poly Bottle
Potency Inclusive between 845μg/mg
Water 2% Max.
Color White
Quantity 25 g
Merck Index 15, 583
Formula Weight 371.39
Chemical Name or Material Ampicillin Sodium Salt
Show More Show Less
compliance-icons
Product Identifier
  • Ampicillin Sodium Salt
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .