Läs mer
Ampiroxicam, MedChemExpress
Produktkod. 16483387
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16483387 50 mg
16417368 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16483387 Leverantör Medchem Express Leverantörsnummer HY17484
MedChemExpress Ampiroxicam(CP65703) is a nonselective cyclooxygenase inhibitor uesd as anti-inflammatory drug.
Ampiroxicam(CP65703) is a nonselective cyclooxygenase inhibitor uesd as anti-inflammatory drug.
Specifikationer
Kemiskt namn eller material Ampiroxicam
CAS 99464-64-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C20H21N3O7S
Kvantitet 50 mg
Synonym CP 65703
Löslighetsinformation DMSO : 50 mg/mL (111.74 mM; Need ultrasonic)
LEDER O=C(OC(OC(C1=CC=CC=C12)=C(C(NC3=NC=CC=C3)=O)N(C)S2(=O)=O)C)OCC
Molekylvikt (g/mol) 447.46
Formel vikt 447.46
Procent renhet 98.43%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation