Läs mer
MedChemExpress α-Amylase
Produktkod. 16412389
Encompass_Preferred
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
16412389 1 g
1 options
Produktkod. 16412389 Leverantör MedChemExpress Leverantörsnummer HYB2193
MedChemExpress α-Amylase is a hydrolase enzyme that catalyses the hydrolysis of internal α-1, 4-glycosidic linkages in starch to yield products like glucose and maltose.
Specifikationer
CAS 9000-90-2
Innehåll och lagring Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Produkttyp Natural Products
Kvantitet 1 g
Frakt skick Room temperature
Löslighet DMSO : 1 mg/mL (Need ultrasonic) ∣H2O : 0.67 mg/mL (Need ultrasonic)