missing translation for 'onlineSavingsMsg'
Learn More

Amylase alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12540-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840294

  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

12540-1-AP can recognize both AMY2A and AMY2B.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.
Specifications

Specifications

Amylase alpha
Polyclonal
Unconjugated
AMY2B
Alpha amylase 2B, AMY2, AMY2B, Amylase, alpha, amylase, alpha 2B (pancreatic), Carcinoid alpha amylase
Rabbit
Antigen Affinity Chromatography
RUO
280, 545562
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P19961
AMY2B
Amylase, alpha Fusion Protein Ag3260
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers