Läs mer
Anagliptin, TRC
Produktkod. 30134032
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134032 50 mg
30005077 1 mg
30005473 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134032 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63721050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 739366-20-2
Molekylformel C19H25N7O2
Synonym N-[2-[[2-[(2S)-2-Cyano-1-pyrrolidinyl]-2-oxoethyl]amino]-2-methylpropyl]-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxamide∣N-[2-([2-[(2S)-2-Cyanopyrrolidin-1-yl]-2-oxoethyl]amino)-2-methylpropyl]-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxamide∣Suiny
Specifikationer
Kemiskt namn eller material Anagliptin
Kvantitet 50 mg
Frakt skick Room Temperature
Formel vikt 383.207
MDL-nummer MFCD19443729
Molekylvikt (g/mol) 383.45, 383.46
InChI-nyckel LDXYBEHACFJIEL-HNNXBMFYSA-N
LEDER CC1=NN2C=C(C=NC2=C1)C(=O)NCC(C)(C)NCC(=O)N1CCC[C@H]1C#N
IUPAC-namn N-[2-({2-[(2S)-2-cyanopyrrolidin-1-yl]-2-oxoethyl}amino)-2-methylpropyl]-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxamide