Läs mer
Anagrelide Hydrochloride, TRC
Produktkod. 30338538
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30338538 5 mg
1 options
Produktkod. 30338538 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6373005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Anagrelide Hydrochloride
CAS 58579-51-4
Molekylformel C10 H7 Cl2 N3 O . Cl H
Kvantitet 5 mg
Synonym 6,7-Dichloro-1,5-dihydroimidazo[2,1-b]quinazolin-2(3H)-one, Imidazo[2,1-b]quinazolin-2(3H)-one, 6,7-dichloro-1,5-dihydro-, monohydrochloride (9CI), 6,7-Dichloro-1,5-dihydroimidazo[2,1-b]quinazolin-2(3H)-one Hydrochloride, Agrelin, Agrylin, Anagrelide hydrochloride, BL 4162A, BMY 26538-01
InChI formel InChI=1S/C10H7Cl2N3O.ClH/c11-6-1-2-7-5(9(6)12)3-15-4-8(16)14-10(15)13-7;/h1-2H,3-4H2,(H,13,14,16);1H
LEDER Cl.Clc1ccc2N=C3NC(=O)CN3Cc2c1Cl
IUPAC-namn 6,7-dichloro-3,5-dihydro-1H-imidazo[2,1-b]quinazolin-2-one;hydrochloride
Molekylvikt (g/mol) 292.55
Formel vikt 290.9733