Learn More
Analyte Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 8140-A
Additional Details : Weight : 0.00100kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Azinphos-Methyl | 86-50-0 | 100μg/mL |
Chlorpyrifos | 2921-88-2 | 100μg/mL |
Coumaphos | 56-72-4 | 100μg/mL |
Demeton O+S | 8065-48-3 | 100μg/mL |
Diazinon | 333-41-5 | 100μg/mL |
Dichlorvos | 62-73-7 | 100μg/mL |
Disulfoton | 298-04-4 | 100μg/mL |
Fenchlorphos | 299-84-3 | 100μg/mL |
Fensulfothion | 115-90-2 | 100μg/mL |
Fenthion | 55-38-9 | 100μg/mL |
Methyl parathion | 298-00-0 | 100μg/mL |
Mevinphos | 7786-34-7 | 100μg/mL |
Phorate | 298-02-2 | 100μg/mL |
Prophos | 13194-48-4 | 100μg/mL |
Sulprofos | 35400-43-2 | 100μg/mL |
Tetrachlorvinphos | 961-11-5 | 100μg/mL |
Tokuthion/Prothiophos | 34643-46-4 | 100μg/mL |
Tributylphosphoro-trithioite | 150-50-5 | 100μg/mL |
Trichloronate | 327-98-0 | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Analyte Mix | |
2mL amber prescored glass ampule | |
Hexane:Acetone | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
SW-846 Method 8140 | |
For US EPA Method 8140 | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Safety and Handling
- Analyte Mix
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : n-hexane, acetone
Product intended for laboratory use only