Läs mer
Analytes B for ICP, 11 Components, Honeywell Fluka™
Produktkod. 16626083 Handla allt Honeywell Fluka Produkter
Change view
Kvantitet:
100 ml
Förpackningsstorlek:
100ml
Produktkod. Kvantitet unitSize
16626083 100 ml 100ml
1 options
Produktkod. 16626083 Leverantör Honeywell Fluka Leverantörsnummer E2156100ML
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Analytes B for ICP
Densitet 1.07 g/cm3
pH <2
Kvantitet 100 ml
Kvalitet Standards for ICP and ICP-MS