Learn More
ANAPC5 Mouse anti-Human, Unconjugated, Clone: 2A7G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67348-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The ANAPC5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ANAPC5. This antibody recognizes human, mouse, rat antigen. The ANAPC5 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ANAPC5 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
2A7G5 | |
Unconjugated | |
150 μL | |
51433 | |
IgG1 |