Läs mer
ABEBA™ Anatom Black/Red Safety Shoes S3 ESD
Storlek:
36
37
38
39
40
41
42
43
44
45
46
47
48
Förpackningsstorlek:
1 par
Beskrivning
- Comfortable, robust and durable shoes
- Comply to EN ISO 20345: 2012, S3, SRC
- ESD dissipative according to EN ISO 61340
- Slip resistant PU double density outsole
- Black/red water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable active comfort insole
Specifikationer
Specifikationer
| Färg | Black, Red |
| Material | Cordura |
| Storlek | 44 |
| Tåmaterial | Steel |
| Certifieringar/efterlevnad (Europa) | EN ISO 20345: 2012 S3 SRC, EN ISO 61340 ESD class 1 |
| Kön | Men's |
| Ankel stil | Low-cut |
| Vattentät | Water Resistant |
| Produkttyp | S3 SRC ESD |