Läs mer
ABEBA™ Anatom Green/Yellow Safety Shoes S3 ESD
Storlek:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Förpackningsstorlek:
1 par
Beskrivning
- Comfortable, robust, durable shoes
- Comply to EN ISO 20345: 2012, S3, SRC
- ESD dissipative according to EN 61340 class climatic 1
- Slip resistant PU double density outsole
- Green/yellow water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable active comfort insole
Specifikationer
Specifikationer
| Färg | Green, Yellow |
| Material | Cordura |
| Storlek | 51 |
| Tåmaterial | Steel |
| Certifieringar/efterlevnad (Europa) | EN ISO 20345: 2012 S3 SRC, ESD class 1 according EN 61340 |
| Kön | Men's |
| Ankel stil | Low-cut |
| Vattentät | Water Resistant |
| Produkttyp | S1P SRC ESD |