Läs mer
(+)-Anatoxin A Hydrochloride, TRC
Produktkod. 30330348
Change view
Kvantitet:
0.5 mg
Förpackningsstorlek:
0.5mg
Produktkod. Kvantitet unitSize
30330348 0.5 mg 0.5mg
1 options
Produktkod. 30330348 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6533600.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-Anatoxin A Hydrochloride
CAS 64314-16-5
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H15 N O . Cl H
Kvantitet 0.5 mg
Synonym Ethanone, 1-(1R,6R)-9-azabicyclo[4.2.1]non-2-en-2-yl-, hydrochloride (9CI), Ethanone, 1-(9-azabicyclo[4.2.1]non-2-en-2-yl)-, hydrochloride, (1R)-, 9-Azabicyclo[4.2.1]nonane, ethanone deriv., (+)-Anatoxin-a hydrochloride, Anatoxin a hydrochloride, Anatoxin a hydrochloride, (+)-Anatoxin A hydrochloride
InChI formel InChI=1S/C10H15NO.ClH/c1-7(12)9-4-2-3-8-5-6-10(9)11-8;/h4,8,10-11H,2-3,5-6H2,1H3;1H/t8-,10-;/m1./s1
LEDER Cl.CC(=O)C1=CCC[C@@H]2CC[C@H]1N2
IUPAC-namn 1-[(1R,6R)-9-azabicyclo[4.2.1]non-4-en-5-yl]ethanone;hydrochloride
Molekylvikt (g/mol) 201.69
Formel vikt 201.092