Learn More
IKA™ R 1330 Anchor Stirrer
Product Code. 10485061 Shop All IKA Products
Change view
Click to view available options
Diameter (Metric) Fan:
45 mm
Diameter (Metric) Shaft:
8 mm
Length (Metric):
350 mm
Speed:
1000 rpm
Unit Size:
Each
Product Code. Diameter (Metric) Fan Diameter (Metric) Shaft Length (Metric) Speed Unit Size
10485061 45 mm 8 mm 350 mm 1000 rpm Each
1 options
Product Code. 10485061 Supplier IKA™ Supplier No. 0002022300
  • ONLINE EXCLUSIVE PRICE
    5855.00 SEK
    5274.64 SEK / Paket med 1
    Save 580.36 SEK
    9% Av
I lager
Overhead, anchor-shaped stirrer element is ideally suited for medium to highly viscous fluids.
  • Tangential flow
  • High shearing rate at edges
  • Minimum deposits on the vessel wall
  • Use at low speeds
  • Use for polymer reactions, even distribution of high mineral contents in liquids
  • Ideal for medium to highly viscous fluids
TRUSTED_SUSTAINABILITY
Specifications
Description R 1330
Diameter (English) Fan 1.8 in.
Diameter (English) Shaft 0.3 in.
Diameter (Metric) Fan 45 mm
Diameter (Metric) Shaft 8 mm
Length (English) 13.8 in.
Length (Metric) 350 mm
Speed 1000 rpm
Type Anchor Impeller