Läs mer
Anecortave, TRC
Produktkod. 17948604
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17948604 5 mg
17919284 25 mg
17939004 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17948604 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6376405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7753-60-8
Molekylformel C23 H30 O5
Molekylvikt (g/mol) 386.48
Synonym 21-(Acetyloxy)-17-hydroxypregna-4,9(11)-diene-3,20-dione, Pregna-4,9(11)-diene-3,20-dione, 17,21-dihydroxy-, 21-acetate (6CI,7CI,8CI), 21-(Acetyloxy)-17-hydroxypregna-4,9(11)-diene-3,20-dione, 2-((10S,13S,17R)-17-Hydroxy-10,13-dimethyl-3-oxo-2,3,6,7,8,10,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl acetate, 21-Acetoxypregna-4,9(11)-dien-17α-ol-3,20-dione, Al 3789, Anecortave, Anecortave acetate, NSC 15475, NSC 24345, Retaane
IUPAC-namn [2-[(8S,10S,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)C3=CC[C@]12C
Specifikationer
Kemiskt namn eller material Anecortave
Analyt- eller komponentnamn Anecortave
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C23H30O5/c1-14(24)28-13-20(26)23(27)11-8-19-17-5-4-15-12-16(25)6-9-21(15,2)18(17)7-10-22(19,23)3/h7,12,17,19,27H,4-6,8-11,13H2,1-3H3/t17-,19+,21+,22+,23+/m1/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 5 mg
Formel vikt 386.2093
Procent renhet >95%