Läs mer
Angiotensin (1-7) xTFA salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Angiotensin (1-7) xTFA salt |
| CAS | 51833-78-4 |
| Molekylformel | C41H62N12O11 . xTFA |
| Kvantitet | 10 mg |
| Synonym | 5-L-Isoleucine-1-7-angiotensin II, 1-7-Human angiotensin, 1-7-Human angiotensin 1, 1-7-Human angiotensin 2, 1-7-Human angiotensin I, 1-7-Human angiotensin II, 1-7-[Ile5]-angiotensin, 115: PN: WO2006032674 SEQID: 115 claimed sequence, 11: PN: WO0212471 SEQID: 18 unclaimed sequence, 11: PN: WO2010028845 SEQID: 9 claimed sequence, 18: PN: US6610497 SEQID: 18 unclaimed sequence, 18: PN: US6884771 SEQID: 18 unclaimed sequence, 18: PN: US6989363 SEQID: 18 unclaimed sequence, 1: PN: US20140205631 SEQID: 4 claimed sequence, 1: PN: US20140296143 SEQID: 1 claimed protein, 1: PN: US20160016996 SEQID: 2 claimed protein, 1: PN: US8633158 SEQID: 1 claimed protein, 1: PN: WO2010028845 SEQID: 1 claimed protein, 1: PN: WO2012021578 SEQID: 1 claimed protein, 1: PN: WO2013032784 SEQID: 1 claimed protein, 1: PN: WO2013090833 SEQID: 1 claimed sequence, 1: PN: WO2013119870 SEQID: 1 claimed sequence, 1: PN: WO2013158959 SEQID: 1 claimed protein, 1: PN: WO2014190152 SEQID: 1 claimed protein, 1: PN: WO2015054005 SEQID: 1 claimed protein, 1: PN: WO2016112247 SEQID: 1 claimed protein, 1: PN: WO2017066552 SEQID: 1 claimed protein, 28: PN: US20160160263 SEQID: 67 claimed protein, 2: PN: WO2014021942 SEQID: 4 claimed protein, 39: PN: US20090227507 SEQID: 1 unclaimed protein, 3: PN: WO0144270 SEQID: 4 claimed protein, 3: PN: WO2008089532 PAGE: 19 unclaimed sequence, 3: PN: WO2011053789 SEQID: 3 claimed protein, 3: PN: WO2013158628 SEQID: 3 unclaimed protein, 3: PN: WO2013172956 SEQID: 4 unclaimed protein, 3: PN: WO2013182237 SEQID: 3 claimed protein, 453: PN: WO0069900 SEQID: 639 unclaimed sequence, 4: PN: WO0056345 SEQID: 4 claimed sequence, 4: PN: WO0143761 SEQID: 4 claimed protein, 4: PN: WO0155176 SEQID: 4 claimed protein, 4: PN: WO0198325 SEQID: 4 claimed protein, 4: PN: WO02087504 SEQID: 4 claimed protein, 4: PN: WO03039434 PAGE: 7 unclaimed sequence, 5-Ile-angiotensin-(1-7), 529: PN: US20090175821 SEQID: 639 claimed protein, 5: PN: US20030130196 SEQID: 4 claimed protein, 6: PN: WO0018899 PAGE: 18 unclaimed sequence, 8: PN: WO03072059 SEQID: 1 claimed protein, Human angiotensin 1-(1-7), Human angiotensin 2-(1-7), Human angiotensin I-(1-7), Human angiotensin II-(1-7), Human angiotensin-(1-7), TXA 127, [Ile5]angiotensin I (1-7) |
| InChI formel | InChI=1S/C41H62N12O11/c1-5-22(4)33(38(61)50-29(17-24-19-45-20-47-24)39(62)53-15-7-9-30(53)40(63)64)52-36(59)28(16-23-10-12-25(54)13-11-23)49-37(60)32(21(2)3)51-35(58)27(8-6-14-46-41(43)44)48-34(57)26(42)18-31(55)56/h10-13,19-22,26-30,32-33,54H,5-9,14-18,42H2,1-4H3,(H,45,47)(H,48,57)(H,49,60)(H,50,61)(H,51,58)(H,52,59)(H,55,56)(H,63,64)(H4,43,44,46)/t22-,26-,27-,28-,29-,30-,32-,33-/m0/s1 |
| LEDER | CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(=N)N)NC(=O)[C@@H](N)CC(=O)O)C(C)C)C(=O)N[C@@H](Cc2cnc[nH]2)C(=O)N3CCC[C@H]3C(=O)O |
| IUPAC-namn | (2S)-1-[(2S)-2-[[(2S,3S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-amino-3-carboxypropanoyl]amino]-5-(diaminomethylideneamino)pentanoyl]amino]-3-methylbutanoyl]amino]-3-(4-hydroxyphenyl)propanoyl]amino]-3-methylpentanoyl]amino]-3-(1H-imidazol-5-yl)propanoyl]pyrrolidine-2-carboxylic acid |
| Molekylvikt (g/mol) | 899 |
| Formel vikt | 898.4661 |