Learn More
Hettich Lab Technology™ Swing-Out Rotor with Carriers
Hettich Lab Technology™ Swing-Out Rotor with Carriers is suitable for performing rate-zonal and isopycnic centrifugations
1810.00 SEK - 8840.00 SEK
Product Code | Brand | Capacity | Max. RCF | Max. Speed | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Max. RCF | Max. Speed | Price | Quantity & Availability | |||
10286674
|
Hettich Lab Technology™
1025 |
4 x 15 mL | 1,077 x g | 3,000 rpm |
1810.00 SEK
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10597861
|
Hettich Lab Technology™
1619 |
15 x 15 mL | 2,701 x g | 4,000 rpm |
8840.00 SEK
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10410333
|
Hettich Lab Technology™
1617 |
8 x 50 mL | 3,857 x g | 5,000 rpm |
8310.00 SEK
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Complete with carriers
- Convenient and easy to use
- Built to withstand long-term, repeated use
Specifications
Swing-out Rotor | |
Carriers |