Learn More
Hettich Lab Technology™ Swing-Out Rotor with Carriers
Hettich Lab Technology™ Swing-Out Rotor with Carriers is suitable for performing rate-zonal and isopycnic centrifugations
1810.00 SEK - 8840.00 SEK
Specifications
Type Swing-out Rotor
Includes Carriers
View More Specs
Products 3
Product Code Brand Capacity Max. RCF Max. Speed Price Quantity & Availability  
Product Code Brand Capacity Max. RCF Max. Speed Price Quantity & Availability  
10286674
View Documents Promotion Details
Hettich Lab Technology™
1025
4 x 15 mL 1,077 x g 3,000 rpm
1810.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
10597861
View Documents Promotion Details
Hettich Lab Technology™
1619
15 x 15 mL 2,701 x g 4,000 rpm
8840.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
10410333
View Documents Promotion Details
Hettich Lab Technology™
1617
8 x 50 mL 3,857 x g 5,000 rpm
8310.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
  • Complete with carriers
  • Convenient and easy to use
  • Built to withstand long-term, repeated use
Specifications
Specifications
Swing-out Rotor
Carriers