missing translation for 'onlineSavingsMsg'
Learn More

ANGPTL8 Mouse anti-Human, Clone: 2H9F9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66641-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878705

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.
Specifications

Specifications

ANGPTL8
2H9F9
Unconjugated
ANGPTL8
Liquid
ANGPTL8
ANGPTL8 Fusion Protein Ag20708
Protein G
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6UXH0
Angiopoietin-like protein 8, ANGPTL8, Betatrophin, C19orf80, Lipasin, LOC55908, PRO1185, PVPA599, td26
Mouse
IgG1
150 μL
-20°C
Primary
55908
Videos
SDS
Documents
Certificates
Special Offers

Special Offers