Läs mer
Anhydro Dihydro Artemisinin, TRC
Produktkod. 30133973
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133973 25 mg
30007830 100 mg
30004666 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63787525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82596-30-3
Molekylformel C15 H22 O4
Molekylvikt (g/mol) 266.33
Synonym (3R,5aS,6R,8aS,12R,12aR)-3,4,5,5a,6,7,8,8a-Octahydro-3,6,9-trimethyl--3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin, 3, 2-Epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin, 3,4,5,5a,6,7,8,8a-octahydro-3,6,9-trimethyl-, [3R-(3α,5aβ,6β,8aβ,12β,12aR*)]-, (+)-9,10-Anhydrodehydroartemisinin, 9,10-Anhydro-10-deoxoartemisinin, 9,10-Anhydrodehydroartemisinin, Anhydrodihydroartemisinin, Anhydrodihydroartemisnin
IUPAC-namn (3R,5aS,6R,8aS,12R,12aR)-3,4,5,5a,6,7,8,8a-Octahydro-3,6,9-trimethyl--3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin
LEDER C[C@@H]1CC[C@H]2C(=CO[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23OO4)C
Specifikationer
Kemiskt namn eller material 9,10-Anhydrodehydroartemisinin
Analyt- eller komponentnamn 9,10-Anhydrodehydroartemisinin
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C15H22O4/c1-9-4-5-11-10(2)8-16-13-15(11)12(9)6-7-14(3,17-13)18-19-15/h8-9,11-13H,4-7H2,1-3H3/t9-,11+,12+,13-,14-,15+/m1/s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 25 mg
Formel vikt 266.1518
Procent renhet >95%