Läs mer
Anhydro Galanthamine, TRC
Produktkod. 30331594
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30331594 25 mg
1 options
Produktkod. 30331594 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA64150025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Anhydro Galanthamine
CAS 664995-65-7
Molekylformel C17 H19 N O2
Kvantitet 25 mg
Synonym 4aH-Benzofuro[3a,3,2-ef][2]benzazepine, 9,10,11,12-tetrahydro-3-methoxy-11-methyl-, (4aS,8aS)-, (4aS,8aS)-9,10,11,12-Tetrahydro-3-methoxy-11-methyl-4aH-benzofuro[3a,3,2-ef][2]benzazepine, Anhydrogalanthamine, R 116937, Anhydrogalantamine, (4aS,8aS)-9,10,11,12-Tetrahydro-3-methoxy-11-methyl-6H-benzofuro[3a,3,2-ef][2]benzazepine, Tetrahydrogalantamine, Didehydrodeoxygalantamine
InChI formel InChI=1S/C17H19NO2/c1-18-10-9-17-8-4-3-5-14(17)20-16-13(19-2)7-6-12(11-18)15(16)17/h3-8,14H,9-11H2,1-2H3/t14-,17-/m0/s1
LEDER COc1ccc2CN(C)CC[C@@]34C=CC=C[C@@H]3Oc1c24
IUPAC-namn (1S,12S)-9-methoxy-4-methyl-11-oxa-4-azatetracyclo[8.6.1.01,12.06,17]heptadeca-6(17),7,9,13,15-pentaene
Molekylvikt (g/mol) 269.34
Formel vikt 269.1416