Learn More
Aniline-13C6, TRC
Product Code. 30133443
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133443 250 mg 250mg
30007773 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133443 Supplier Toronto Research Chemicals Supplier No. TRCA662478250MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100849-37-4
Molecular Formula 13C6 H7 N
Molecular Weight (g/mol) 99.08
Synonym Aniline 13C6, Benzenamine-13C6 (9CI), Aniline-13C6, Benzenamine-1,2,3,4,5,6-13C6
IUPAC Name (1,2,3,4,5,6-^{13}C_{6})cyclohexatrienamine
SMILES N[13c]1[13cH][13cH][13cH][13cH][13cH]1
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2/i1+1,2+1,3+1,4+1,5+1,6+1
Formula Weight 99.078
Percent Purity >95%
Chemical Name or Material Aniline 13C6
Grade HPLC