missing translation for 'onlineSavingsMsg'
Learn More

Aniline Blue Diammonium Salt (Technical Grade), TRC

Product Code. 30343844
Change view
Click to view available options
Kvantitet:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30343844 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343844 Supplier Toronto Research Chemicals Supplier No. TRCA66250350G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Aniline Blue Diammonium Salt (Technical Grade)
CAS 66687-07-8
Molekylformel C37H29N3O9S3 . 2NH3
Kvantitet 50 g
Synonym Acid blue 93:1, C.I. 427801, C.I. Acid Blue 93:1, Duasyn Acid Blue SN, Ink Blue R, Simacid Soluble Blue 8B, [[4-[Bis[4-[(sulfophenyl)amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]amino]-benzenesulfonic Acid Diammonium Salt
InChI formel InChI=1S/C37H29N3O9S3.H3N/c41-50(42,43)34-19-13-31(14-20-34)38-28-7-1-25(2-8-28)37(26-3-9-29(10-4-26)39-32-15-21-35(22-16-32)51(44,45)46)27-5-11-30(12-6-27)40-33-17-23-36(24-18-33)52(47,48)49;/h1-24,38-39H,(H,41,42,43)(H,44,45,46)(H,47,48,49);1H3
LEDER N.OS(=O)(=O)c1ccc(Nc2ccc(cc2)C(=C3C=CC(=Nc4ccc(cc4)S(=O)(=O)O)C=C3)c5ccc(Nc6ccc(cc6)S(=O)(=O)O)cc5)cc1
IUPAC-namn azane;4-[4-[[4-(4-sulfoanilino)phenyl]-[4-(4-sulfophenyl)iminocyclohexa-2,5-dien-1-ylidene]methyl]anilino]benzenesulfonic acid
Molekylvikt (g/mol) 772.86
Formel vikt 772.1331
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.