Läs mer
Aniline D-Glucuronide Sodium Salt, TRC
Produktkod. 30342703
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30342703 5 mg
1 options
Produktkod. 30342703 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6625005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Aniline D-Glucuronide Sodium Salt
CAS 116866-42-3
Molekylformel C12 H14 N O6 . Na
Kvantitet 5 mg
Synonym Glucopyranuronic acid, 1-anilino-1-deoxy-, sodium salt (9CI), D-Glucopyranuronic acid, 1-deoxy-1-(phenylamino)-, monosodium salt, D- (6CI)
InChI formel InChI=1S/C12H15NO6.Na/c14-7-8(15)10(12(17)18)19-11(9(7)16)13-6-4-2-1-3-5-6;/h1-5,7-11,13-16H,(H,17,18);/q;+1/p-1/t7-,8-,9+,10-,11?;/m0./s1
LEDER O[C@@H]1[C@@H](O)C(Nc2ccccc2)O[C@@H]([C@H]1O)C(=O)O[Na]
IUPAC-namn [(2S,3S,4S,5R)-6-anilino-3,4,5-trihydroxy-tetrahydropyran-2-carbonyl]oxysodium
Molekylvikt (g/mol) 291.23
Formel vikt 291.0719