missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anion Eluent Concentrate, Ion Pair Chromatography, Fisher Chemical
Click to view available options
Quantity:
100 mL
Packaging:
HDPE plastic bottle
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
Color | Colorless |
MDL Number | 282123 |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'