missing translation for 'onlineSavingsMsg'
Learn More

Anisole-d8, TRC

Product Code. 30338014
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30338014 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30338014 Supplier Toronto Research Chemicals Supplier No. TRCA673903250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Anisole-d8
CAS 54887-54-6
Molekylformel C7 D8 O
Kvantitet 250 mg
Synonym 6-(Methoxy-d3)-benzene-1,2,3,4,5-d5, Methoxy-d3-benzene-d5, Perdeuteroanisole
InChI formel InChI=1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D
LEDER [2H]c1c([2H])c([2H])c(OC([2H])([2H])[2H])c([2H])c1[2H]
IUPAC-namn 1,2,3,4,5-pentadeuterio-6-(trideuteriomethoxy)benzene
Molekylvikt (g/mol) 116.19
Formel vikt 116.1077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.