Läs mer
Anisotropine Methylbromide, TRC
Produktkod. 30134970
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134970 100 mg
30132484 1 g
30006811 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134970 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA673960100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80-50-2
Molekylformel C17 H32 N O2 . Br
Molekylvikt (g/mol) 362.35
Synonym 8-Azoniabicyclo[3.2.1]octane, 8,8-dimethyl-3-[(1-oxo-2-propylpentyl)oxy]-, bromide, (3-endo)-, 1αH,5αH-Tropanium, 3α-hydroxy-8-methyl-, bromide, 2-propylvalerate (8CI), 8-Azoniabicyclo[3.2.1]octane, 8,8-dimethyl-3-[(1-oxo-2-propylpentyl)oxy]-, bromide, endo-, 8-Methyltropinium bromide, 2-propylvalerate (6CI,7CI), 8-Methyl-3-(2-propylpentanoyloxy)tropinium bromide, 8-Methyltropinium bromide 2-propylpentanoate, Anisotropine methobromide, Anisotropine methylbromide, Lytispasm, Octatropine methylbromide, Valpin, Valpin 50, Vapin
IUPAC-namn [(1R,5S)-8,8-dimethyl-8-azoniabicyclo[3.2.1]octan-3-yl] 2-propylpentanoate;bromide
LEDER [Br-].CCCC(CCC)C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)[N+]2(C)C
Specifikationer
Kemiskt namn eller material Anisotropine Methylbromide
Kvantitet 100 mg
InChI formel InChI=1S/C17H32NO2.BrH/c1-5-7-13(8-6-2)17(19)20-16-11-14-9-10-15(12-16)18(14,3)4;/h13-16H,5-12H2,1-4H3;1H/q+1;/p-1/t14-,15+,16+;
Formel vikt 361.1616