Learn More
Abnova™ ANK1 (Human) Recombinant Protein
Human ANK1 full-length ORF ( AAH30957, 1 a.a. - 155 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000286-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MWTFVTQLLVTLVLLSFFLVSCQNVMHIVRGSLCFVLKHIHQELDKELGESEDLSDDEETISTRVVRRRVFLKGNEFQNIPGEQVTEEQFTDEQGNIVTKKIIRKVVRQIDLSSADAAQEHEEVELRGSGLQPDLIEGRKGAQIVKRASLKRGKQ
Specifications
AAH30957 | |
ankyrin 1, erythrocytic | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANK1 | |
GST |
286 | |
Wheat germ expression system | |
10 μg | |
ANK, SPH1, SPH2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |