missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24000-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.Specifications
ANKRD13C | |
Polyclonal | |
Unconjugated | |
ANKRD13C | |
ANKRD13C, ankyrin repeat domain 13C, RP4 677H15.5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
433667, 81573 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UX43, Q8N6S4 | |
Ankrd13c | |
ANKRD13C Fusion Protein Ag21202 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |